Transfer Isolators, 3 Gloves, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
4001791 4001792 4001793
76557-588EA 4807.29 CAD
76557-588 76557-722 76557-568
Transfer Isolators, 3 Gloves, Rheo Flexibles (Division of Rheo Engineering)
Glove Boxes and Glove Bags
The Rheo transfer isolator is a single use isolator that provides high containment operator protection during the handling of potentially hazardous powders. This quick deployment solution can be installed quickly and can be disposed of when finished eliminating the cleaning costs associated with traditional stainless steel isolators.
  • Constructed from RheoFlex 200 series film polyurethane material
  • FDA compliant
  • 21 CFR 177.1680 compliant
  • Clear and matte finish
  • Anti-static film
  • BSE/TSE compliant
The bungee strap system and the general flexibility of soft-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable