VWR® Plus Laboratory Chest Freezer with Manual Defrost (15 cu. ft.)
Supplier: VWR
VWR®
VWR-MFP-15-C
76580-192EA 6991.44 CAD
76580-192
VWR® Plus Laboratory Chest Freezer with Manual Defrost (15 cu. ft.)
Freezers Laboratory / General Purpose Freezers
These VWR® Plus Series manual defrost chest freezers offer premium cold storage with superior temperature control.
  • Microprocessor temperature controller
  • Digital temperature display and alarm module
  • Audible, visual, senor error, and power failure alarms
  • Removable baskets
  • LED interior lighting
  • Natural hydrocarbon refrigerants
These manual defrost laboratory chest freezers offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, and precise temperature monitoring through microprocessor control. The Plus Series line includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Material: White powder coat
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.