VWR® Standard Laboratory Refrigerators Solid Door with Natural Refrigerants
Supplier: Horizon Scientific
VWR-HC-RFP-14 76580-188 76580-190
76580-186EA 6994.14 CAD
76580-186 76580-188 76580-190
VWR® Standard Laboratory Refrigerators Solid Door with Natural Refrigerants
Refrigerators
These VWR® Standard Series laboratory refrigerators provide quality cold storage at an economical pricepoint.
  • Microprocessor temperature controller
  • Digital temperature display and alarm module
  • Audible, visual, senor error, and power failure alarms
  • Alarm validation and alarm mute with ring back
  • Keyed door lock
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation,More Product Information
Order Now
SPECIFICATIONS
Cat. No. 76580-186 76580-188 76580-190
Environmentally Preferable Energy Efficient Energy Efficient Energy Efficient
Defrost Cycle Cycle Cycle
Shelves/drawers 5 adjustable shelves 5 adjustable shelves 5 adjustable shelves
Door type 1 solid door, right hinged 1 solid door, right hinged 1 solid door, right hinged
Electrical 115 V, 60 Hz, 3 A, ⅕ HP 115 V, 60 Hz, 3 A, ¼ HP 115 V, 60 Hz, 3 A, ¼ HP
Weight 91 kg (200 lbs.) 109 kg (240 lbs.) 118 kg (260 lbs.)
Environmental certification Uses Natural Hydrocarbon refrigerants Uses Natural Hydrocarbon refrigerants Uses Natural Hydrocarbon refrigerants