VWR® Plus Laboratory Refrigerator/Freezer Combo Units with Natural Refrigerants
Supplier: VWR International
76580-176 76580-178 76580-180 76580-182 76580-184
76580-176EA 8945.74 CAD
76580-176 76580-178 76580-180 76580-182 76580-184
VWR® Plus Laboratory Refrigerator/Freezer Combo Units with Natural Refrigerants
Refrigerators
These VWR® Plus Series laboratory combo units offer premium cold storage with superior temperature control.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual alarm systems
  • LED interior lighting, and adjustable shelving
  • Keyed door lock
  • Natural, hydrocarbon refrigerant
These efficient refrigerator and freezer combo units provide quality cold storage in laboratory settings. These multi-functional units offer a cold-storage solution for a variety of laboratory needs. All models are equipped with temperature alarms, keyed door locks, interior shelving, probe access and precise temperature monitoring through microprocessor control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: UL/C-UL listed.
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Caution: *Compatible with refrigerator compartment only.