VWR® Plus Speciality Laboratory Refrigerators with Glass Door
Supplier: Horizon Scientific
VWR-HC-19 76580-156 76580-160 76580-164
76580-152EA 12376.33 CAD
76580-152 76580-156 76580-160 76580-164
VWR® Plus Speciality Laboratory Refrigerators with Glass Door
Refrigerators
These VWR® Plus Series laboratory refrigerators offer premium cold storage with superior temperature control.
  • Microprocessor temperature controller
  • Digital temperature display and alarm module
  • Audible, visual, senor error, and power failure alarms
  • Alarm validation and alarm mute with ring back
  • Keyed door lock
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These speciality glass door laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, probe access, and precise temperature monitoring through microprocessor control. The Plus Series line includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Material: White powder coat
Certifications: UL/C-UL listed.
*Select models ENERGY STAR certified: 76580-152, 76580-156
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.