VWR® Performance Speciality Laboratory Refrigerators with Glass Door
Supplier: VWR International
76580-166 76580-154 76580-158 76580-162
76580-166EA 27940.12 CAD
76580-166 76580-154 76580-158 76580-162
VWR® Performance Speciality Laboratory Refrigerators with Glass Door
Refrigerators
These VWR® Performance Series speciality laboratory refrigerators offer the highest level features with advanced data logging and superior temperature control.
  • Touch screen digital temperature display
  • Extensive alarm systems
  • Temperature data logging and USB port for data transfer
  • Microprocessor temperature controller
  • LED interior lighting and adjustable shelving
  • Keyed door lock
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These specialty glass door laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, probe access, and precise temperature monitoring through microprocessor control. The Performance line showcases the most elite features including state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Material: White powder coat
Certifications: UL/C-UL listed.
*Select models ENERGY STAR certified: 76580-154, 76580-158
Ordering information: All units come with a three year parts and labor warranty, plus an additional two year compressor parts warranty.