Rapid Response™ Fentanyl Test Strip Kits (Liquid/Powder)
Supplier: BTNX INC.
FYL-18S7-5 FYL-18S7-10 FYL-18S7-5
76539-042CT 3257.72 CAD
76539-042 CA77675-050 CA76539-042
Rapid Response™ Fentanyl Test Strip Kits (Liquid/Powder)
Drug Tests
The Rapid Response™ Fentanyl (FYL) forensic test kit is a lateral flow chromatographic immunoassay for the qualitative detection of fentanyl in liquid and powder substances at the cut-off concentration of 200 ng/ml.
  • Sample: Liquid/Powder
  • Detects fentanyl at a cut-off concentration of 200 ng/ml or higher
  • Full kit with all materials required to test drug samples
  • Results interpretation guide printed on each pouch
  • Easy to interpret results with built-in procedural controls
  • Individually packaged strips
  • Sensitive to fentanyl and analogues: Carfentanil, Butyryl
More Product Information