Michels Solution , Statlab
Supplier: StatLab
MS0507 MS0735 MS-32
76548-150CS 348.83 CAD
76548-150 76548-152 76548-154
Michels Solution , Statlab
Stains Microscopy and staining reagents
Michels Solution is used to preserve and transport specimens for examination in immunofluorescence studies.
  • While not a fixative, specimens can be held at room temperature for five days in transport medium before processing
  • This simple salt solution maintains pH, but does not kill most pathogens
  • Specimens received in transport medium should be washed in three changes of Michels Wash Solution (10 minutes for each wash)
  • Not suitable for FISH studies
Part of the StatLab line of formalin alternatives.
Manufactured in the USA