Ceramic Magnetic Hotplate Stirrers, AREC Series, VELP Scientifica

Supplier: VELP SCIENTIFIC INC.
F20510551 F20510554 SA20510550 SA20510554 SB20510550 SB20510554 SC20510554
76559-348EA 1079.79 CAD
76559-348 76559-340 76559-350 76559-344 76559-346 76559-354 76559-342
Ceramic Magnetic Hotplate Stirrers, AREC Series, VELP Scientifica
Stirrers Magnetic Hotplate Stirrers
AREC and AREC.X hotplate stirrers are designed to provide safety, reliability and the highest heating and stirring performance. Their resistant ceramic square top ensures temperature homogeneity, thermal efficiency and a durable heating performance over time.

  • Temperature setting resolution: 5 °C, except AREC Connect which is 1 °C
  • Different models are available to adapt to laboratory requirements of accuracy, precision and automation
  • Models have a techno polymer housing
  • Stir volumes up to 20 L liters (H₂O), thanks to the powerful motor and magnetic coupling
  • These digital models equipped with a bright, easy to read digital display with white LED allowing operators to easily monitor temperature and stirring speed set
  • The extremely resistant ceramic plate is easy to clean and resistant to acids, bases and solvents
  • Monophase PCM motor with electronic regulation torque compensation

AREC - The basic solution for everyday applications: The AREC is the safe and reliable hot plate stirrer with a ceramic top guaranteeing excellent heat transfer and stirring performance for basic laboratory applications.

AREC.X - The optimal solution for accurate thermoregulation: The AREC.X is designed for accurate thermoregulation offering outstanding stirring and heating performance, superior safety features and ease of use. The ideal solution for laboratories requiring precise control of the medium's temperature. Connection for PT100, available with choice of PT100 or VTF thermoregulator packages.
• Thermoregulation accuracy: ±1 °C ( with PT100)/±0.5 °C ( with VTF)

AREC Connect – Innovative unit for data-rich laboratory processes: Precise thermoregulation and powerful stirring for microbiological, pharmaceutical, chemical and cosmetic laboratories requiring accuracy and efficiency. Integral timer, programming options and Wi-Fi connectivity (ERMES connection) to monitor and control multiple laboratory reactions through VELP Ermes cloud platform. Connection for PT100, probe included in the package.
• Speed setting resolution: 5 min⁻¹
• Intemitant mode and Autoreverse: (5 s to 99 min 59 s)
• Brushless motor with SpeedServo™ torque compensation
• Thermoregulation accuracy: ±1 °C (PT100)
• Lock function and acoustic warning signal

Delivery information: AREC Connect, AREC.X and AREC are supplied with a VELP magnetic stirring bar.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR