VWR® Plus Series Solid Door Laboratory Refrigerators
Supplier: VWR International
76579-122 76579-126 76579-102 76579-106 76579-110 76579-114 76579-118
76579-122EA 20507.39 CAD
76579-122 76579-126 76579-102 76579-106 76579-110 76579-114 76579-118
VWR® Plus Series Solid Door Laboratory Refrigerators
Refrigerators
These VWR® Plus Series laboratory refrigerators offer premium cold storage solutions with superior temperature control.
  • Microprocessor temperature controller
  • Digital temperature display and alarm module
  • Audible, visual, senor error, and power failure alarms
  • Keyed door lock, LED interior lighting, and adjustable shelving
  • Alarm validation and alarm mute with ring back
  • Natural, hydrocarbon refrigerant
  • ENERGY STAR certified*
These VWR® refrigerators are designed with premium features for laboratory use. They feature a series of alarms for product protection and remote alarm contacts. Units employ forced draft air circulation for superior temperature control. These refrigerators utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Material: White powder coat
Certifications: ETL Listed.
*Select models ENERGY STAR certified: 76579-106, 76579-110, 76579-114, 76579-122
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.