VWR® Flammable Storage Laboratory Refrigerators Standard Series with Natural Refrigerants
Supplier: VWR International
76579-148 76579-150 76579-152
76579-148EA 7769.84 CAD
76579-148 76579-150 76579-152
VWR® Flammable Storage Laboratory Refrigerators Standard Series with Natural Refrigerants
Refrigerators
These laboratory refrigerators are designed for the safe storage of flammable substances.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual temperature alarms
  • Probe access port
  • Keyed door lock
  • Natural, hydrocarbon refrigerant
  • Utilizes ATEX and non-sparking components for safety
  • ENERGY STAR certified*
These laboratory refrigerators safely store and protect flammable substances. The interior cabinets of these units utilize ATEX and non-sparking components for safety and peace of mind.
All units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
Material: White powder coat
Certifications: ETL Listed.
*Select models ENERGY STAR certified: 76579-150
Ordering information: All units come with a two year parts and labor warranty.