SureSTART™ Glass Crimp Top Headspace Vials, Level 3 High Performance Applications
Supplier: Thermo Scientific
6PCV20-1FT 6PCV20-1R 6PCV20-1RF 6PCV20-1RT 6PCV6-1F 6PCV6-1FF 6PCV6-1FT 6PCV10-1F 6PCV10-1FT 6PCV20-1F 6PCV20-1FF 6PCV10-V1
TS6PCV20-1FTCS 826.66 CAD
TS6PCV20-1FT TS6PCV20-1R TS6PCV20-1RF TS6PCV20-1RT TS6PCV6-1F TS6PCV6-1FF TS6PCV6-1FT TS6PCV10-1F TS6PCV10-1FF TS6PCV10-1FT TS6PCV20-1F TS6PCV20-1FF TS6PCV10-V1
SureSTART™ Glass Crimp Top Headspace Vials, Level 3 High Performance Applications
Vials
Use Thermo Scientific™ SureSTART™ glass crimp top headspace vials, performance level 3, for your volatile gas analyzes.
  • Round base vials are compatible with most autosamplers and more easily handled by robotic arms that lift the vial from the tray
  • Flat base vials maximize heating efficiency in manual headspace sampling and are required for use in some instrument models
  • Identified by three waves on the vial
  • Convenient write-on patch
  • Suitable for GC and mass spectrometry (MS) applications
  • 100% traceable through the manufacturing process
Available in 6, 1More Product Information