SureSTART™ Total Recovery Glass Screw Top 1.5 ml Microvials for <2 ml Samples, Level 3 High Performance Applications
Supplier: Thermo Scientific
6PSV9-TR1F 6PSV9-TR1T 6PSV9-TR1 6PSV9-CHV1 6PSV9-SV1
TS6PSV9-TR1FCS 21892.94 CAD
TS6PSV9-TR1F TS6PSV9-TR1T TS6PSV9-TR1 TS6PSV9-CHV1 TS6PSV9-SV1
SureSTART™ Total Recovery Glass Screw Top 1.5 ml Microvials for <2 ml Samples, Level 3 High Performance Applications
Vials
Choose Thermo Scientific™ SureSTART™ 1.5 ml total recovery glass screw top microvials, performance Level 3, when you need to maximize the injection volume for <2 ml samples.
  • Specification Certified to ensure their physical dimensions and autosampler compatibility
  • Meets all requirements of US, EU, JPN Pharmacopeia
  • Seal with 9 mm screw caps and septa
  • 100% traceable through the manufacturing process
  • All Thermo Scientific SureSTART vials are dimensionally verified
  • Inspected by automated optical and gauge inspection to ensure critical dimensions and
More Product Information