SureSTART™ Glass Screw Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PSV9-03FIVAP 6PSV9-03FIVAPF 6PSV9-03FIVAPT 6PSV9-03FIVP 6PSV9-03FIVPF 6PSV9-03FIVPT
TS6PSV903FIVAPPK 173.6 CAD
TS6PSV903FIVAP TS6PSV93FIVAPF TS6PSV9-3FIVAP TS6PSV9-03FIVP TS6PSV903FIVPF TS6PSV903FIVPT
SureSTART™ Glass Screw Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 0.3 ml glass screw top microvials, performance level 3, when you need to maximize the injection volume for <2 ml samples.
This 12×32 mm microvials has a fixed insert that provides a total usable volume of 0.25 ml and a residual volume of 4 µl. Available in clear and amber glass, this is a performance level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes.