SureSTART™ Glass Crimp Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PCV1103FIVAP 6PCV1103FIVAPF 6PCV1103FIVAPT 6PCV11-03FIVP 6PCV11-03FIVPF 6PCV11-03FIVPT
TS6PCV11-3FIVAPPK 205.21 CAD
TS6PCV11-3FIVAP TS6PCV1103FIVA TS6PCV113FIVAP TS6PCV11-3FIVP TS6PCV113FIVPF TS6PCV1103FIVP
SureSTART™ Glass Crimp Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 0.3 ml glass crimp top microvials, performance level 3, when you need to maximize the injection volume for <2 ml samples.
This 12×32 mm microvials has a fixed insert, providing a total usable volume of 0.25 ml and a residual volume of 4 µl. Available in clear and amber glass, this is a performance level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes. They are specification certified to ensure physical dimensions and autosampler compatibility.More Product Information