SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PSV9-1PSS 6PSV9-1PSSF 6PSV9-1PSST 6PSV9-1PT 6PSV9-2PSS 6PSV9-2PSSF 6PSV9-2PSST 6PSV9-S1P 6PSV9-1P 6PSV9-1PF
TS6PSV9-1PSSPK 41.04 CAD
TS6PSV9-1PSS TS6PSV9-1PSSF TS6PSV9-1PSST TS6PSV9-1PT TS6PSV9-2PSS TS6PSV9-2PSSF TS6PSV9-2PSST TS6PSV9-S1P TS6PSV9-1P TS6PSV9-1PF
SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml GOLD-grade clear glass screw and crimp top vials, performance level 3, when analyzing analytes such as polar compounds that strongly adsorb on the glass surface or are sensitive to sample pH changes.
Level 3 vials are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes. This offering includes Thermo Scientific™ SureStop™ vials, which, when used with 9 mm AVCS™ screw caps, ensure an optimal seal every time. The SureStop vial-AVCS cap system is specially designed to prevent screw thread vial failures such as cap tilt, cap deformities, dislodged septa, or septum push through, thus preventing injection failures and sample evaporation.