SureSTART™ Glass Crimp Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PCV11-1P 6PCV11-1PF 6PCV11-1PT
TS6PCV11-1PPK 33.06 CAD
TS6PCV11-1P TS6PCV11-1PF TS6PCV11-1PT
SureSTART™ Glass Crimp Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials, performance level 3, for high performance applications, when sensitivity is a must. Our level 3 vials are our highest performance vials, designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding GC-MS. These 2 ml clear glass crimp vials are specification certified to ensure their physical dimensions and autosampler compatibility. Use 11 mm crimp caps and septa to seal these vials. Seal and remove your crimp caps using Thermo Scientific manual and electronic crimpers and decrimpers. These vials meet all requirements of US, EU, JPN Pharmacopeia. Buy in bulk for greater savings.
  • Performance level 2
  • SureSTART vials & cap closures for high throughput applications
  • Get the performance you need to deliver high quality data with an uninterrupted workflow
  • Our performance level 2 range has very low metal content and extractables and leachables, so that you have the assurance that each product conforms to the same high standards you are working with to meet to your regulatory requirements
  • If you need products that support routine analyzes, our performance level 2 products are here for you select the products with two waves
About Thermo Scientific SureSTART Vials, Caps, and Kits
Thermo Scientific SureSTART vials and caps are available in three performance levels. Now you can choose the products that best suit your analytical, performance, and budget needs.
Level 1 vials and caps are for everyday applications and are suitable for QA/QC applications using core detectors.
Level 2 vials, caps, and kits are for high throughput applications. These vials have very low metal content, extractables, and leachables to deliver high quality data with an uninterrupted workflow.
Level 3 vials, caps, and kits are for high performance applications. Use level 3 for your most valuable samples (and analytes prone to glass adsorption) to deliver consistent, reproducible results for your most demanding analyzes.
Certifications: Specification certified.
All Thermo Scientific SureSTART vials are dimensionally verified.
Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances.
Uniform and defect-free.
Lot traceable.
100% traceable through the manufacturing process.
Certificate of Conformance available by lot number upon request.