SureSTART™ Polyspring Inserts for 2 ml Vials, Level 2 High-Throughput, Thermo Scientific
Supplier: Thermo Scientific
6EME03CPPSP 6EME03CPPSPF 6EME03CPPSPT
TS6EME03CPPSPPK 71.11 CAD
TS6EME03CPPSP TS6EME03CPPSPF TS6EME03CPPSPT
SureSTART™ Polyspring Inserts for 2 ml Vials, Level 2 High-Throughput, Thermo Scientific
Vials
Use Thermo Scientific™ polyspring plastic inserts with 2 ml screw, crimp, or snap vials to maximize injection volume when analyzing <2 ml samples.
The integral polyspring at the bottom of the insert centers the insert in the vials, ensures a good seal with the vials cap, and provides a cushion against needle contact. These plastic inserts have a small total volume capacity of <400 μl and a minimum residual volume of 2 µl. When placed and sealed inside a 2 ml vials, microsamples can be used in conventional autosampler trays. These inserts are performance level 2, which means they were designed to ensure high quality data with an uninterrupted workflow in high-throughput applications. They are specification certified to ensure physical dimensions.