SureSTART™ Crimp Caps, 11 mm, Level 3 High Performance Applications
Supplier: Thermo Scientific
6PCC11ST101 6PCC11TST 6PCCMST101X 6ACC11NR1Y 6PCC11ST101X
TS6PCC11ST101EA 103.03 CAD
TS6PCC11ST101 TS6PCC11TST TS6PCCMST101X TS6ACC11NR1Y TS6PCC11ST101X
SureSTART™ Crimp Caps, 11 mm, Level 3 High Performance Applications
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 11 mm crimp caps with crimp vials that have an 11 mm opening, including SureSTART 2 ml glass crimp vials and glass crimp microvials for <2 ml samples (Level 3). Select level 3 products for ultimate performance and sensitivity from methods, while maintaining the highest levels of sample security and integrity.
Choose from aluminum and magnetic steel caps with all-purpose silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Use Thermo Scientific™ manual or electronic crimpers and decappers for easy application andMore Product Information