SureSTART™ Polypropylene Snap Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
6ERV11-03PPC 6ERV11-03PPCF 6ERV11-03PPCT 6ERV11-08PPC 6ERV11-08PPCF 6ERV11-08PPCT 6ERV11-08PPFB 6ERV11-08PPFBF 6ERV11-08PPFBT
TS6ERV11-03PPCPK 46.31 CAD
TS6ERV11-03PPC TS6ERV1103PPCF TS6ERV1103PPCT TS6ERV11-08PPC TS6ERV1108PPCF TS6ERV1108PPCT TS6ERV-118PPFB TS6ERV1108PPFB TS6ERV11-8PPFB
SureSTART™ Polypropylene Snap Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Vials
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml polypropylene snap top microvials for everyday chromatography analysis of <2 ml samples. They provide an affordable choice and can be used with all HPLC/UHPLC instrument types.
These clear plastic 2 ml snap top vials are specification certified to ensure autosampler compatible physical dimensions. Seal these snap top vials with 11 mm snap caps and septa.
Order Now
SPECIFICATIONS
Environmentally Preferable