SureSTART™ Polypropylene Screw Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6ESV9-04PP 6ESV9-1PP 6ESV9-1PPF 6ESV9-04PPT 6ESV9-1PPT 6ESV9-04PPF
TS6ESV9-04PPFCS 1909.08 CAD
TS6ESV9-04PPF TS6ESV9-04PPT TS6ESV9-1PP TS6ESV9-1PPF TS6ESV9-04PP TS6ESV9-1PPT
SureSTART™ Polypropylene Screw Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Vials
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml polypropylene screw top microvials for everyday chromatography analysis of <2 ml samples. They provide an affordable choice and can be used with all HPLC/UHPLC instrument types.
These clear plastic 2 ml screw top microvials are specification certified to ensure autosampler compatible physical dimensions. Seal these screw top vials with 9 mm screw caps and septa.