SureSTART™ Glass Crimp Top Vials, 2 ml, evel 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
CHCV11-20PF CHCV11-20PT CHCV11-10PF CHCV11-10PT
TSCHCV11-20PFCS 1427.72 CAD
TSCHCV11-20PF TSCHCV11-20PT TSCHCV11-10PF TSCHCV11-10PT
SureSTART™ Glass Crimp Top Vials, 2 ml, evel 1 Everyday Analysis, Thermo Scientific
Vials
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials for everyday chromatography analysis. They provide an affordable choice and can be used with all GC and HPLC/UHPLC instrument types.
These 2 ml glass crimp top vials are specification certified to ensure autosampler compatible physical dimensions. Choose from clear and amber glass. The vials meet all requirements of US, EU, and JPN pharmacopia. Seal these crimp vials with 11 mm aluminum crimp caps and septa using a manual or electronic crimper tool.