SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6ARC11ST1 6ARC11ST1F 6ARC11ST1R 6ARC11ST1RF 6ARC11ST1RT 6ARC11ST1T 6ARC11NRT1 6ARC11NRT1F 6ARC11NRT1R 6ARC11NRT1RT 6ARC11NRT1T
TS6ARC11ST1EA 49.83 CAD
TS6ARC11ST1 TS6ARC11ST1F TS6ARC11ST1R TS6ARC11ST1RF TS6ARC11ST1RT TS6ARC11ST1T TS6ARC11NRT1 TS6ARC11NRT1F TS6ARC11NRT1R TS6ARC11NRT1RF TS6ARC11NRT1RT TS6ARC11NRT1T
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Closures Chromatography Vial Closures
Choose Thermo Scientific™ SureSTART™ 2 ml glass snap vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems.
Use level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. Available in clear and amber glass, snap top vials have very low metal content, extractables, and leachables and conform to high standards so that you can to meet your regulatory requirements. They are specification certifiedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable