SureSTART™ Caps for 20 ml Glass Crimp Top Headspace Vials, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
CHCC20-2F CHCC20-2MT CHCC20-2T CHCC20-1F CHCC20-1MT CHCC20-1T
TSCHCC20-2FCS 4149.79 CAD
TSCHCC20-2F TSCHCC20-2MF TSCHCC20-2MT TSCHCC20-2T TSCHCC20-1F TSCHCC20-1MF TSCHCC20-1MT TSCHCC20-1T
SureSTART™ Caps for 20 ml Glass Crimp Top Headspace Vials, Level 1 Everyday Analysis, Thermo Scientific
Closures Chromatography Vial Closures
Use performance level 1 Thermo Scientific™ SureSTART™ 20 ml glass crimp top vials for everyday chromatography analysis. They provide an affordable choice and can be used with all headspace GC instrument types.
These headspace vials are made of superior quality 1st hydrolytic class glass (Type 1) and have a thicker glass wall to withstand higher temperatures and internal pressures for the analysis of volatile gas in the vial’s headspace. The vials are specification certified to ensure autosampler compatible physical dimensions, and meet all requirements of US, EU, and JPN pharmacopia. SealMore Product Information