SureSTART™ Caps for 2 ml Glass Crimp Top Vials, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
CHCC11-10F CHCC11-10T CHCC11-30F CHCC11-30T
TSCHCC11-10FCS 960.02 CAD
TSCHCC11-10F TSCHCC11-10T TSCHCC11-30F TSCHCC11-30T
SureSTART™ Caps for 2 ml Glass Crimp Top Vials, Level 1 Everyday Analysis, Thermo Scientific
Closures Chromatography Vial Closures
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials for everyday chromatography analysis. They provide an affordable choice and can be used with all GC and HPLC/UHPLC instrument types.
These 2 ml glass crimp top vials are specification certified to ensure autosampler compatible physical dimensions. Choose from clear and amber glass. The vials meet all requirements of US, EU, and JPN pharmacopia. Seal these crimp vials with 11 mm aluminum crimp caps and septa using a manual or electronic crimper tool.