SureSTART™ EPA Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6AK8CCT 6AK12CCT 6AK16ACT 6AK16CCT 6AK20ACTAS 6AK20CCTAS 6AK40ACT 6AK40AOTAS 6AK40CCT 6AK40COTAS 6AK4ACT 6AK4CCT 6AK8ACT 6AK12ACT
TS6AK8CCTPK 226.64 CAD
TS6AK8CCT TS6AK12CCT TS6AK16ACT TS6AK16CCT TS6AK20ACTAS TS6AK20CCTAS TS6AK40ACT TS6AK40AOTAS TS6AK40CCT TS6AK40COTAS TS6AK4ACT TS6AK4CCT TS6AK8ACT TS6AK12ACT
SureSTART™ EPA Certified Screw Vial and Cap Kits, Level 2 High-throughput Applications, Thermo Scientific
Vials
Ensure that your environmental analyzes meet government regulations with our level 2 Thermo Scientific™ SureSTART™ EPA certified screw vial and cap kits. Select level 2 products for high throughput applications, when robustness and reproducibility are key.
Choose from a variety of kits containing screw vials in clear or amber glass and open or closed top polypropylene caps with silicone/PTFE or polyethylene foam/PTFE septa designed to reduce autosampler needle issues.