SureSTART™ Glass Screw Top Headspace Vials, 20 ml, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6ASV20-1 6ASV20-1F 6ASV20-1T 6ASV20-2 6ASV20-2T
TS6ASV20-1PK 113.76 CAD
TS6ASV20-1 TS6ASV20-1F TS6ASV20-1T TS6ASV20-2 TS6ASV20-2F TS6ASV20-2T
SureSTART™ Glass Screw Top Headspace Vials, 20 ml, Level 2 High-Throughput Applications, Thermo Scientific
Vials
Use Thermo Scientific™ SureSTART™ 20 ml glass screw top headspace vials in your high-throughput volatile gas analyzes. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC and single or triple quadrupole MS systems.
These headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyzes. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements. These vials are specification certified to ensure their physical dimensions and autosampler compatibility.