SureSTART™ Glass Screw Top Vials, 4 ml, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6ASV13-1P 6ASV13-2P
TS6ASV13-1PPK 46.31 CAD
TS6ASV13-1P TS6ASV13-2P
SureSTART™ Glass Screw Top Vials, 4 ml, Level 2 High-Throughput Applications, Thermo Scientific
Vials
Use Thermo Scientific™ SureSTART™ 4 ml glass screw top headspace vials, performance level 2, in your high-throughput volatile gas analyzes. Our screw top headspace vials are a novelty in a crimp-neck dominated field.
They provide a ready-to-use, convenient solution that does not require crimping or decrimping tools. These are performance level 2 vials that ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems. The headspace vials are made to withstand higher temperatures and internalMore Product Information