SureSTART™ Glass Crimp Top Headspace Vials, 20 ml, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6ACV20-1PE 6ACV20-1PEF 6ACV20-1PET 6ACV20-1R 6ACV20-1RF 6ACV20-1RT 6ACV20-2PE 6ACV20-2PET
TS6ACV20-1PEPK 82.35 CAD
TS6ACV20-1PE TS6ACV20-1PEF TS6ACV20-1PET TS6ACV20-1R TS6ACV20-1RF TS6ACV20-1RT TS6ACV20-2PE TS6ACV20-2PEF TS6ACV20-2PET
SureSTART™ Glass Crimp Top Headspace Vials, 20 ml, Level 2 High-Throughput Applications, Thermo Scientific
Vials
Use Thermo Scientific™ SureSTART™ 20 ml glass crimp top headspace vials in your high-throughput volatile gas analyzes. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC and single or triple quadrupole MS systems.
These headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyzes. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements. These vials are specification certified to ensure theirMore Product Information