SureSTART™ Glass Crimp Top Headspace Vials, 10 ml, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6ACV10-1RT 6ACV10-2R 6ACV10-2RF 6ACV10-2RT 6ACV10-1R 6ACV10-1RF
TS6ACV10-1RTCS 761.97 CAD
TS6ACV10-1RT TS6ACV10-2R TS6ACV10-2RF TS6ACV10-2RT TS6ACV10-1R TS6ACV10-1RF
SureSTART™ Glass Crimp Top Headspace Vials, 10 ml, Level 2 High-Throughput Applications, Thermo Scientific
Vials
Use Thermo Scientific™ SureSTART™ 10 ml glass crimp top headspace vials in your high-throughput volatile gas analyzes. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC and single or triple quadrupole MS systems.
These headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyzes. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements. These vials are specification certified to ensure their physical dimensions and autosampler compatibility.