SureSTART™ Glass Screw Top Vials, 2 ml, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6ASV9-2PT 6ASV9-S2P 6ASV9-1P 6ASV9-1PF 6ASV9-1PT 6ASV9-2P 6ASV9-2PF
TS6ASV9-2PTCS 454.36 CAD
TS6ASV9-2PT TS6ASV9-S2P TS6ASV9-1P TS6ASV9-1PF TS6ASV9-1PT TS6ASV9-2P TS6ASV9-2PF
SureSTART™ Glass Screw Top Vials, 2 ml, Level 2 High-Throughput Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml glass screw top vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems. Use level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. Available in clear and amber glass, these 2 ml screw top vials have very low metal content, extractables, and leachables and conform to high standards so that you can to meet your regulatory requirements. They are specification certified to ensure their physical dimensions and autosampler compatibility.
  • Performance level, level 2
  • SureSTART vials & cap closures for high throughput applications
  • Get the performance you need to deliver high quality data with an uninterrupted workflow
  • Our performance level 2 range has very low metal content and extractables and leachables, so that you have the assurance that each product conforms to the same high standards you are working with to meet to your regulatory requirements
  • If you need products that support routine analyzes, our performance level 2 products are here for you select the products with two waves.
About Thermo Scientific SureSTART vials, caps, and kits, Thermo Scientific SureSTART vials and caps are available in three performance levels. Now you can choose the products that best suit your analytical, performance, and budget needs. Level 1 vials and caps are for everyday applications and are suitable for QA/QC applications using core detectors. Level 2 vials, caps, and kits are for high throughput applications. These vials have very low metal content, extractables, and leachables to deliver high quality data with an uninterrupted workflow. Level 3 vials, caps, and kits are for high performance applications. Use level 3 for your most valuable samples (and analytes prone to glass adsorption) to deliver consistent, reproducible results for your most demanding analyzes.
Certifications: Specification certified. All Thermo Scientific SureSTART vials are dimensionally verified. Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances. Uniform and defect-free. Lot traceable. 100% traceable through the manufacturing process. Certificate of conformance available by lot number upon request.