SureSTART™ Stoppers, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
6A20BS 6A20BSF
TS6A20BSEA 41.81 CAD
TS6A20BS TS6A20BSF TS6A20BST
SureSTART™ Stoppers, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Stoppers Rubber Stoppers
Use Thermo Scientific™ SureSTART™ 20 mm stoppers with crimp vials that have a 20 mm opening and a secure push fit, including our SureSTART 20 mm crimp headspace vials (level 2).
Select level 2 products for high throughput applications, when robustness and reproducibility are key. Choose from various pack sizes of low permeability, 3 mm thick bromobutyl rubber stoppers designed to provide an effective alternative to crimp caps.