SureSTART™ Precision Screw Caps, 18 mm, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PMSC18-ST2 6PMSC18-ST2F 6PMSC18-ST2T 6PMSC18-STH 6PMSC18-STHT
TS6PMSC18-ST2PK 174.12 CAD
TS6PMSC18-ST2 TS6PMSC18-ST2F TS6PMSC18-ST2T TS6PMSC18-STH TS6PMSC18-STHF TS6PMSC18-STHT
SureSTART™ Precision Screw Caps, 18 mm, Level 3 High Performance Applications, Thermo Scientific
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 18 mm precision screw caps with screw vials that have an 18 mm opening, including our SureSTART 10 ml and 20 ml screw top headspace vials (level 3). Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity.
Choose from magnetic steel caps with all-purpose silicone/PTFE septa designed to reduce autosampler needle issues. Select high temperature septa to allow for heating to over 250 °C without excessive septum softening or the release of volatile vapors.