SureSTART™ Electronic Hand-Held Crimpers and Decrimpers, Level 2 High Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
60180-ECRHB 60180-ECR11 60180-ECR20 60180-EDCB20 60180-EDCB13 60180-ECR13 60180-EDCB11 60180-ECR8
TS60180-ECR20EA 1884.31 CAD
TS60180-ECR20 TS60180-ECR11 TS60180-EDCB13 TS60180-ECRHB TS60180-EDCB11 TS60180-EDCB20 TS60180-ECR13 TS60180-ECR8
SureSTART™ Electronic Hand-Held Crimpers and Decrimpers, Level 2 High Throughput Applications, Thermo Scientific
Crimpers, Cappers and Decappers
Use Thermo Scientific™ electronic hand-held vial crimpers and decrimpers to attach or remove crimp caps from vials, respectively.
These hand-held, high-power crimpers and decrimpers are available in 8, 11, 13, and 20 mm sizes and allow you to seal or remove crimp caps with a simple, one-handed push of a button. Use for crimping of vials with aluminum, pressure-release, and bi-metal caps. The electronic crimper's message window conveniently provides all-important information at one glance. This device is performance level 3, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.