SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PK1655 6PKG592W
TS6PK1655CS 2269.12 CAD
TS6PK1655 TS6PKG592W
SureSTART™ Specification Certified Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Vials
Meet your certification and documentation needs with our level 3 Thermo Scientific™ SureSTART™ specification certified screw vial and cap kits.
Select Level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity. Choose from a variety of kits containing short thread screw vials in glass or TPX and polypropylene caps with silicone/PTFE septa designed to reduce autosampler needle issues. Bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps for effective seals,More Product Information