SureSTART™ MSCERT Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PMSCK540W 6PMSCK541W 6PMSCK601W 6PMSCK611W 6PMSCK641W 6PMSCK651W 6PMSCK534W 6PMSCK535W
TS6PMSCK540WPK 156.82 CAD
TS6PMSCK540W TS6PMSCK541W TS6PMSCK601W TS6PMSCK611W TS6PMSCK641W TS6PMSCK651W TS6PMSCK534W TS6PMSCK535W
SureSTART™ MSCERT Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Vials
Meet your certification and documentation needs with our level 3 Thermo Scientific™ SureSTART™ MSCERT screw vial and cap kits. Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity.
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass and polypropylene caps with silicone/PTFE septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps forMore Product Information