SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PME05F1 6PME03C1SP 6PME03C1SPF 6PME03C1SPT 6PME03C1SSP 6PME03C1SSPF 6PME03C1SSPT 6PME04C1 6PME04C1F 6PME04C1T
TS6PME05F1EA 129.54 CAD
TS6PME05F1 TS6PME03C1SP TS6PME03C1SPF TS6PME03C1SPT TS6PME03C1SSP TS6PME03C1SSPF TS6PME03C1SSPT TS6PME04C1 TS6PME04C1F TS6PME04C1T
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Vials
Use Thermo Scientific™ SureSTART™ performance level 3 inserts with 2 ml vials when analysing <2 ml samples to maximise the injection volume. These glass inserts have a small total volume capacity of <400 μl, with a minimum residual volume of 2 µl.
When placed and sealed inside a 2 ml vial, microsamples can be used in conventional autosampler trays. Choose from polyspring conical, pulled point conical, or flat bottom insert formats. Conical inserts are most effective with residual volumes of 2 to 4 μl. These inserts are performance level 3, which means they are designed to protectMore Product Information