SureSTART™ High Recovery Glass Snap Top Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PRV11-15V 6PRV11-15VT 6PRV11-S1V
TS6PRV11-15VPK 165.88 CAD
TS6PRV11-15V TS6PRV11-15VF TS6PRV11-15VT TS6PRV11-S1V
SureSTART™ High Recovery Glass Snap Top Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ high recovery glass snap top microvials, performance level 3, to maximise injection volume when analysing <2 ml samples.
With outer dimensions of 12×32 mm, these clear and amber glass microvials have an inner V-shaped bottom to enable a relatively high total usable volume and a residual volume of 4 µl. This is a performance level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.