SureSTART™ Glass Snap Top Microvials, 0.3 ml for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PRV11-03FIVP 6PRV11-03FIVPF 6PRV11-03FIVPT
TS6PRV11-3FIVPPK 170.34 CAD
TS6PRV11-3FIVP TS6PRV1103FIVP TS6PRV1103FIVT
SureSTART™ Glass Snap Top Microvials, 0.3 ml for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ glass snap top microvials, performance level 3, to maximise the injection volume when analysing <2 ml samples.
With outer dimensions of 12×32 mm, this clear or amber glass microvial has a conical-shaped, fixed inner insert to enable a usable volume of 0.25 ml and a residual volume of 4 µl. This is a performance level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. They are specification certified to ensure physical dimensions and autosamplerMore Product Information