SureSTART™ Snap Caps, 11 mm, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
6PRC11ST1 6PRC11ST101 6PRC11ST101F 6PRC11ST101R 6PRC11ST101RF 6PRC11ST101RT 6PRC11ST101T 6PRC11ST1F 6PRC11ST1R 6PRC11ST1RF 6PRC11ST1RT 6PRC11ST1T 6PRC11STS1 6PRC11STS1F 6PRC11STS1R 6PRC11STS1RF 6PRC11STS1RT 6PRC11STS1T 6PRC11STS1X 6PRC11STS1XF 6PRC11STS1XT
TS6PRC11ST1PK 66.69 CAD
TS6PRC11ST1 TS6PRC11ST101 TS6PRC11ST101F TS6PRC11ST101R TS6PRC11ST10RF TS6PRC11ST10RT TS6PRC11ST101T TS6PRC11ST1F TS6PRC11ST1R TS6PRC11ST1RF TS6PRC11ST1RT TS6PRC11ST1T TS6PRC11STS1 TS6PRC11STS1F TS6PRC11STS1R TS6PRC11STS1RF TS6PRC11STS1RT TS6PRC11STS1T TS6PRC11STS1X TS6PRC11STS1XF TS6PRC11STS1XT
SureSTART™ Snap Caps, 11 mm, Level 3 High Performance Applications, Thermo Scientific
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 11 mm Snap caps with snap vials that have an 11 mm opening, including our SureSTART 2 ml glass snap Vials, 1.5 ml total recovery glass snap vials, high recovery glass snap vials, and glass snap micro vials for <2 ml samples (level 3).
Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity. Choose from soft polyethylene caps with double snap functionality that guarantees a tight seal.
These snap caps include all-purpose silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Pre-slit septa options are available for those who want septa that prevent vial-overfilling issues and re-seal well enough to avoid sample evaporation.