ChromCap™ 100 HPLC Cap Assembly, Foxx Life Sciences
Supplier: Foxx Life Sciences
450-0320-FLS 450-0321-FLS 450-0322-FLS 450-0620-FLS 450-0621-FLS 450-0622-FLS 450-0623-FLS 450-0624-FLS
76552-482EA 240.7 CAD
76552-482 76552-484 76552-486 76552-488 76552-490 76552-492 76552-494 76552-496
ChromCap™ 100 HPLC Cap Assembly, Foxx Life Sciences
Closures Bottle Closures
Foxx Life Sciences ChromCap™ 100 used in upstream caps and bottle system, offer the safest and most ergonomic lab safety solution in the industry.
  • Patented VersaCap® technologyprovides the flexibility of interchangeable connectors
  • To prevent spills and vapors fromleaking, tube fittings secure tubes tocontainer to create a close-loopsystem
  • Plugs included for all unused ports
Foxx Life Sciences ChromCap™ 100 used in upstream caps and bottle system, offer the safest and most ergonomic lab safety solution in the industry. This system comes with patented VersaCap®, a Class VI PTFE manifold, chemically resistant viton o-ring. The sealed VersaCap® design that Foxx offers eliminates tubes twisting as you thread the cap and eliminates the tubes from sliding in and out of the container.
Each assembly comes with either a 38-430 mm cap or a patented GL 45 cap and manifold assembly, high quality fittings for 1×3.2 mm (¹/₈") Ext.Ø tubing, 1×1.6 mm (¹/₁₆") Ext.Ø tubing and 1×plugs of 6.4 mm (¹/₄") for blocking of unused ports.