Ring Light Package LED3000 for S9 Series, Leica Microsystems
Supplier: Leica Microsystems
8121491 8121491
CA76575-536EA 1052.3 CAD
76575-536 CA76575-536
Ring Light Package LED3000 for S9 Series, Leica Microsystems
Lights Ring Lights
Ring lights provide bright and uniform illumination of samples.
  • Longer lifetimes (25000 to 50000 hours)
  • Consume less power
  • Natural color temperature
  • Cold light source (less heat release)
  • Compact design
The LED3000 Ring Light is compatible with the S9 1.0× optic lens and the 0.63× supplementary lens. It will not work with the 1.6× lens. Additional diffusors and polarized light sets are available, which reduces glare and highlights.
Delivery information: The LED3000 ring light package comes with the LED3000 ring light, power supply and cable.