EZBio® Media Bottles, Square Storage Bottles, with and without Caps, Foxx Life Sciences
Supplier: Foxx Life Sciences
137-1114-FLS 137-1115-FLS 137-1116-FLS 137-1117-FLS 137-2213-FLS 137-3214-FLS 137-4214-FLS 137-4215-FLS 137-5213-FLS 137-6513-FLS 138-1111-FLS 138-1113-FLS 138-3213-FLS 138-4213-FLS 138-5213-FLS 138-6513-FLS
76552-432PK 288.62 CAD
76552-432 76552-434 76552-436 76552-438 76552-440 76552-442 76552-444 76552-446 76552-448 76552-450 76552-452 76552-454 76552-456 76552-458 76552-460 76552-462 76552-464 76552-466 76552-468
EZBio® Media Bottles, Square Storage Bottles, with and without Caps, Foxx Life Sciences
Bottles Plastic Bottles
Foxx Life Sciences offers the widest range of EZBio® Single Use Media Bottles on the market, with a variety of sizes, cap options, and materials available.
  • Media bottles are available in five different sizes: 60 ml, 125 ml, 250 ml, 500ml, 1 L, 2 L
  • Made entirely of 100% USP Class VI components from the United States
  • Gamma Irradiated in the United States
  • EZBio® Single Use Media Bottles are compatible with the EZBio® cap assemblies
With a leakproof design and revolutionary VersaCap® technology, Foxx offers the most cost effective product on the market. EZBio® Bottles can be used for sampling, sorting media, biologicals, and many other aqueous solutions.
Certifications: USP Class VI material.