Soft-Grip Spatulas, United Scientific Supplies
Supplier: United Scientific Supplies
SSSG08 SSSG03 SSSG04 SSSG05 SSSG06
470339-310EA 31.9 CAD
470339-310 470339-686 470340-124 470339-482 470339-240
Soft-Grip Spatulas, United Scientific Supplies
Spatulas
Comfort grip spatulas with soft silicone.
  • Laboratory spatula with stainless steel blade
  • Sturdy polypropylene handle covered in a soft silicone grip for more comfortable use
  • Latex-free
  • Autoclavable
Laboratory spatula with 3-inch blade features a sturdy polypropylene handle covered in a soft silicone grip for more comfortable use. Blade is made from stainless steel with a flexible, rounded end.