CO₂ Incubators with Hot Air Sterilization and Humidity Regulation, CBF Series, Binder

Supplier: Binder
9640-0019 9640-0020 9640-0023 9640-0024 9640-0026
76575-494EA 25297.54 CAD
76575-494 76575-496 76575-498 76575-500 76575-502
CO₂ Incubators with Hot Air Sterilization and Humidity Regulation, CBF Series, Binder
Incubators CO₂ Incubators
The CBF series CO₂ incubators are the premium series of CO₂ incubators with humidity control. Units are suitable for all sensitive incubation applications and ensure optimal cell growth in GMP environments. A CBF series CO₂ incubator can be used for complex cultivation experiments or individual environments with hypoxic conditions thanks to the comprehensive range of options and accessories. Units ensure consistent results including in cell-based assays with multiwell plates.

  • Contamination-safe humidification system without water pan
  • Tried-and-tested anti-
More Product Information
Order Now

SPECIFICATIONS

Model CBF 170 CBF 260
Environmentally Preferable
Temperature variation (spatial) ±0,4 °C @ 37 °C ±0,4 °C @ 37 °C
Recovery time after 30 second door opening at 37 °C 6 min 6 min
Convection type Air jacket system Air jacket system
CO₂ recovery time after 30 second door opening at 5 Vol.% CO₂ 5 min 5 min
Shelves supplied (max.) 3 (6) 3 (8)
Max. load per shelf 10 kg 10 kg
Max. chamber workload 66 lbs. (30 kg) 88 lbs. (40 kg)
Int. W×D×H 22.79×19.9×23.64" 24.43×22.65×29.55"
Ext. W×D×H 26.79×28.17×34.28" 29.16×30.93×40.2"
Energy consumption 55 Wh/h at 37 °C 65 Wh/h at 37 °C
Power consumption 1.4 kW 1.5 kW
Sound level 41 dB(A) 41 dB(A)
Temperature range Ambient +6…60 °C Ambient +6…60 °C
Electrical 100/120 V; 1~ 50/60 Hz 100/120 V; 1~ 50/60 Hz
Weight 209 lbs. (95 kg) 276 lbs. (125 kg)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR