CO₂ Incubators with Hot Air Sterilization, CB Series, Binder
Supplier: Binder
9640-0028 9640-0007 9640-0011 9640-0012 9640-0015 9640-0016
76575-504EA 22692.75 CAD
76575-504 76575-482 76575-484 76575-486 76575-488 76575-490 76575-492
CO₂ Incubators with Hot Air Sterilization, CB Series, Binder
Incubators CO₂ Incubators
CO₂ incubators with a built in CO₂ sensor which can be sterilized. Units offer optimum growth conditions for cell cultures and meet demanding requirements for best possible cell growth and maximum sample safety. The ANTI.PLENUM design minimizes surfaces due to the seamless, deep-drawn inner chamber without fixtures and featuring an integrated shelf support system. This reduces the risk of contamination, giving less surface area and fewer parts to clean manually, and removes the running costs of consumables. Reliable decontamination is provided by the standard-compliant 180 °C hot air sterilization routine which completely eliminates contaminants and ensures the inner chamber is completely sterilized.
  • Touch-screen controller with color LCD and internal data recording via USB interface
  • CO₂ introduction through Venturi gas mixing nozzle
  • Electronic error self-diagnostic system with visual and acoustic alarms, as well as a potential free contact for central monitoring
  • Independent temperature safety device Class 3.1 (DIN 12880) with visual and audible temperature alarm
  • Ethernet interface for APT-COM™ Multi Management Software
  • Stable pH values due to short recovery times with drift-free infrared CO₂ measurement system
Uniform growth conditions are provided by the VENTAIR™ jacket system, this ensures accurate temperatures, homogeneous temperature distribution, and enables fast recovery times. It also minimizes thermal degradation of cell growth due to occasional door openings, and the Permadry™ double-pan humidification system ensures high humidity and condensate-free interior walls. Units can be stacked using stacking adapter to save space, have a tightly fitted inner glass door made of safety glass and two-point door lock.
Certifications: CE marked
Delivery information: Supplied with perforated stainless steel shelves and Binder test certificate. Calibrations and validations possible. A wide range of options and accessories are available, including partitioned inner glass door and divided shelves), O₂ control, gas tank connection kit for CO₂/O₂/N₂, for details please contact your local Avantor sales office.