Compact Glove Boxes, Yamato
Supplier: Yamato Scientific
SG-828 SG-835 SG-848 SG-860
76557-482EA 26890.78 CAD
76557-482 76557-480 76557-488 76557-484
Compact Glove Boxes, Yamato
Glove Boxes and Glove Bags
Compact, completely sealed glove box with transfer chamber and flat access door for basic isolation and/or containment.
  • High visibility interior, cleanliness, safety, complete containment, portability, and a draft-free atmopshere
  • Completed systems, nothing needs to be added except gas of choice: nitrogen, argon, or other inert gases
  • Installed purging valves are ideal for lowering oxygen and humidity levels
Very useful when working with toxic substances, asbestos, fibers, sewage residue, and harmful liquid vapors. Made in the USA.
Certifications: CE certification.
Order Now
SPECIFICATIONS
Environmentally Preferable