Eisco Micro Bunsen Burner with Flame Stabilizer
Supplier: Wards
CH0093D CH0093E
470343-868EA 75.05 CAD
470343-868 470345-826
Eisco Micro Bunsen Burner with Flame Stabilizer
Heaters Gas Burners
Adjustable orifice and flame stabilizing burner tip.
  • Equipped with a flame stabilizing burner tip and air ports with collar to adjust the flow of air
  • Gas flow is controlled by needle valve fitted above the base permitting easy flame adjustment
  • Suitable for use with LPG (liquid petroleum gas) / butane gas or mixed gas with a heat value of 800 to 1200 BTU
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Aluminum mixing tube is mounted on a circular, nickel plated die casted zinc alloy base
Nickel-plated die casted zinc alloy base, aluminum mixing tube, flame stabilizing burner tip and adjustable air ports. Air supply is controlled by a nickel plated brass sleeve covering ports at the lower end of the mixing tube, and gas flow is controlled by needle valve fitted above the base for easy flame adjustment. Burner measures 3.75" in height and base measures 3" in Ø.