Eisco Glass Aspirator Bottles with Ground Socket Top and Outlet
Supplier: Wards
CH0062D CH0062C CH0062A CH0062B
470344-252EA 535.95 CAD
470344-252 470345-056 470345-272 470345-814
Eisco Glass Aspirator Bottles with Ground Socket Top and Outlet
Bottles Aspirators
Ungraduated glass aspirator bottles have a ground socket sidearm outlet located at the base of the bottle for attaching a compatible stopcock (available separately), and an open, ground socket neck.
  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Superior durability and chemical resistance
  • Ground joints at outlet and neck for attaching stopcock or other compatible glassware (available separately)
Glass aspirator bottles are autoclavable, and offers superior chemical durability and a low temperature gradient. They can withstand high temperatures and thermal shocks.
Eisco borosilicate glass has the following heat tolerances: strain point of 515 °C, annealing point of 565 °C, and softening point of 820 °C.