Eisco Tirrill Burner
Supplier: Wards
CH0095A CH0095B
470345-230EA 96 CAD
470345-230 470345-680
Eisco Tirrill Burner
Heaters Gas Burners
Designed to provide the most precise control of flame.
  • Both air and gas supply can be adjusted, allowing for separate control of flame height and temperature
  • Gas control provided by a thumb knob on the bottom of the burner
  • Adjustable air ports control air supply
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Highly stable octagonal base
An additional level of control is gained when using a Tirrill Burner instead of a standard burner. Burner measures 6.25" tall, and the base measures 3.25" wide.